1-[(Ammoniooxy)methyl]-4-fluorbenzolchlorid

Lieferant: BIONET RESEARCH

Synonyme: O-(4-Fluorobenzyl)hydroxylamine hydrochloride

3F-940.500 3F-940.1 3F-940.5 3F-940.10
BIOT3F-940.500EA 0 EUR
BIOT3F-940.500 BIOT3F-940.1 BIOT3F-940.5 BIOT3F-940.10
1-[(Ammoniooxy)methyl]-4-fluorbenzolchlorid
1-[(Ammoniooxy)methyl]-4-fluorbenzolchlorid
Formel: C₇H₉ClFNO
Molecular Weight: 177.6039 g/mol
MDL: MFCD01114581
CAS-Nummer: 51572-89-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR