Thiazolidin-2,4-dicarbonsäure
Lieferant: BIONET RESEARCH
Synonyme:
Tidiacic, Tetrahydrothiazole-2,4-dicarboxylic acid
3J-939.500
3J-939.1
3J-939.5
3J-939.10
BIOT3J-939.500EA
0
EUR
BIOT3J-939.500
BIOT3J-939.1
BIOT3J-939.5
BIOT3J-939.10
Thiazolidin-2,4-dicarbonsäure
Thiazolidin-2,4-dicarbonsäure
Formel:
C₅H₇NO₄S Molecular Weight: 177.1784 g/mol |
MDL:
MFCD00145399 CAS-Nummer: 30097-06-4 EINECS: 250-048-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...