Thiazolidin-2,4-dicarbonsäure

Lieferant: BIONET RESEARCH

Synonyme: Tidiacic, Tetrahydrothiazole-2,4-dicarboxylic acid

3J-939.500 3J-939.1 3J-939.5 3J-939.10
BIOT3J-939.500EA 0 EUR
BIOT3J-939.500 BIOT3J-939.1 BIOT3J-939.5 BIOT3J-939.10
Thiazolidin-2,4-dicarbonsäure
Thiazolidin-2,4-dicarbonsäure
Formel: C₅H₇NO₄S
Molecular Weight: 177.1784 g/mol
MDL: MFCD00145399
CAS-Nummer: 30097-06-4
EINECS: 250-048-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR