1-(Phenylsulfonyl)-4-piperidincarbonsäure

Lieferant: BIONET RESEARCH

3R-1011.500 3R-1011.1 3R-1011.5 3R-1011.10
BIOT3R-1011.500EA 0 EUR
BIOT3R-1011.500 BIOT3R-1011.1 BIOT3R-1011.5 BIOT3R-1011.10
1-(Phenylsulfonyl)-4-piperidincarbonsäure
1-(Phenylsulfonyl)-4-piperidincarbonsäure
Formel: C₁₂H₁₅NO₄S
MDL: MFCD00723116
CAS-Nummer: 122891-92-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR