1-(Phenylsulfonyl)-4-piperidincarbonsäure
Lieferant: BIONET RESEARCH
3R-1011.500
3R-1011.1
3R-1011.5
3R-1011.10
BIOT3R-1011.500EA
0
EUR
BIOT3R-1011.500
BIOT3R-1011.1
BIOT3R-1011.5
BIOT3R-1011.10
1-(Phenylsulfonyl)-4-piperidincarbonsäure
1-(Phenylsulfonyl)-4-piperidincarbonsäure
Formel:
C₁₂H₁₅NO₄S |
MDL:
MFCD00723116 CAS-Nummer: 122891-92-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...