5-(4-Morpholinyl)-2-nitroanilin

Lieferant: BIONET RESEARCH

3T-0352.500 3T-0352.1 3T-0352.5
BIOT3T-0352.500EA 0 EUR
BIOT3T-0352.500 BIOT3T-0352.1 BIOT3T-0352.5
5-(4-Morpholinyl)-2-nitroanilin
5-(4-Morpholinyl)-2-nitroanilin
Formel: C₁₀H₁₃N₃O₃
MDL: MFCD00511844
CAS-Nummer: 54998-00-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR