4'-Chloracetophenonoxim
Lieferant: BIONET RESEARCH
Synonyme:
1-(4-Chlorophenyl)-1-ethanone oxime
4F-002.500
4F-002.1
4F-002.5
4F-002.10
BIOT4F-002.500EA
0
EUR
BIOT4F-002.500
BIOT4F-002.1
BIOT4F-002.5
BIOT4F-002.10
4'-Chloracetophenonoxim
4'-Chloracetophenonoxim
Formel:
C₈H₈ClNO |
MDL:
MFCD02609517 CAS-Nummer: 1956-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...