4-Amino-1H-indazol
Lieferant: BIONET RESEARCH
4P-101.500
4P-101.1
4P-101.5
4P-101.10
BIOT4P-101.500EA
0
EUR
BIOT4P-101.500
BIOT4P-101.1
BIOT4P-101.5
BIOT4P-101.10
4-Amino-1H-indazol
4-Amino-1H-indazole
Formel:
C₇H₇N₃ Molecular Weight: 133.15 g/mol |
MDL:
MFCD03305710 CAS-Nummer: 41748-71-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...