4-Amino-1H-indazol

Lieferant: BIONET RESEARCH

4P-101.500 4P-101.1 4P-101.5 4P-101.10
BIOT4P-101.500EA 0 EUR
BIOT4P-101.500 BIOT4P-101.1 BIOT4P-101.5 BIOT4P-101.10
4-Amino-1H-indazol
4-Amino-1H-indazole
Formel: C₇H₇N₃
Molecular Weight: 133.15 g/mol
MDL: MFCD03305710
CAS-Nummer: 41748-71-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR