1-((4-Fluorphenyl)sulfonyl)-4-piperidincarbonsäure

Lieferant: BIONET RESEARCH

4R-1333.500 4R-1333.1 4R-1333.5 4R-1333.10
BIOT4R-1333.500EA 0 EUR
BIOT4R-1333.500 BIOT4R-1333.1 BIOT4R-1333.5 BIOT4R-1333.10
1-((4-Fluorphenyl)sulfonyl)-4-piperidincarbonsäure
1-((4-Fluorphenyl)sulfonyl)-4-piperidincarbonsäure
Formel: C₁₂H₁₄FNO₄S
MDL: MFCD02331439
CAS-Nummer: 326907-70-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR