2-Amino-3-brom-5-chlorpyridin

Lieferant: BIONET RESEARCH
Achtung

Synonyme: 3-Brom-5-chlorpyridin-2-amin, 3-Brom-5-chlor-2-pyridylamin

4T-0646.500 4T-0646.1 4T-0646.5 4T-0646.10 KO4T-0646.0001
BIOT4T-0646.500EA 0 EUR
BIOT4T-0646.500 BIOT4T-0646.1 BIOT4T-0646.5 BIOT4T-0646.10 ABCRKO4T-0646.0001
2-Amino-3-brom-5-chlorpyridin
2-Amino-3-brom-5-chlorpyridin
Formel: C₅H₄BrClN₂
Molecular Weight: 207.46 g/mol
MDL: MFCD00955627
CAS-Nummer: 26163-03-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR