2-Hydroxy-4'-methoxyacetophenon

Lieferant: BIONET RESEARCH

Synonyme: α-Hydroxy-4'-methoxyacetophenon

4X-0724.500 4X-0724.1 4X-0724.5 4X-0724.10
BIOT4X-0724.500EA 0 EUR
BIOT4X-0724.500 BIOT4X-0724.1 BIOT4X-0724.5 BIOT4X-0724.10
2-Hydroxy-4'-methoxyacetophenon
2-Hydroxy-4'-methoxyacetophenon
Formel: C₉H₁₀O₃
Molecular Weight: 166.18 g/mol
MDL: MFCD00004668
CAS-Nummer: 4136-21-4
EINECS: 223-955-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR