2-(4-Fluorphenyl)-5-nitropyridin
Lieferant: BIONET RESEARCH
4Y-0704.500
4Y-0704.5
4Y-0704.10
4Y-0704.1
BIOT4Y-0704.500EA
0
EUR
BIOT4Y-0704.500
BIOT4Y-0704.5
BIOT4Y-0704.10
BIOT4Y-0704.1
2-(4-Fluorphenyl)-5-nitropyridin
2-(4-Fluorophenyl)-5-nitropyridine
Formel:
C₁₁H₇FN₂O₂ Molecular Weight: 218.19 g/mol |
MDL:
MFCD06200966 CAS-Nummer: 886361-78-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...