3-Formyl-6-methylchromon

Lieferant: BIONET RESEARCH

Synonyme: 6-Methyl-4-oxo-4H-chromene-3-carbaldehyde

5C-082.500 5C-082.1 5C-082.5 5C-082.10
BIOT5C-082.500EA 0 EUR
BIOT5C-082.500 BIOT5C-082.1 BIOT5C-082.5 BIOT5C-082.10
3-Formyl-6-methylchromon
3-Formyl-6-methylchromon
Formel: C₁₁H₈O₃
Molecular Weight: 188.1794 g/mol
MDL: MFCD00138943
CAS-Nummer: 42059-81-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR