3-Formyl-6-methylchromon
Lieferant: BIONET RESEARCH
Synonyme:
6-Methyl-4-oxo-4H-chromene-3-carbaldehyde
5C-082.500
5C-082.1
5C-082.5
5C-082.10
BIOT5C-082.500EA
0
EUR
BIOT5C-082.500
BIOT5C-082.1
BIOT5C-082.5
BIOT5C-082.10
3-Formyl-6-methylchromon
3-Formyl-6-methylchromon
Formel:
C₁₁H₈O₃ Molecular Weight: 188.1794 g/mol |
MDL:
MFCD00138943 CAS-Nummer: 42059-81-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...