4-[(Ammoniooxy)methyl]-1,2-dichlorbenzolchlorid

Lieferant: BIONET RESEARCH

5E-042.500 5E-042.1 5E-042.5 5E-042.10
BIOT5E-042.500EA 0 EUR
BIOT5E-042.500 BIOT5E-042.1 BIOT5E-042.5 BIOT5E-042.10
4-[(Ammoniooxy)methyl]-1,2-dichlorbenzolchlorid
4-[(Ammoniooxy)methyl]-1,2-dichlorbenzolchlorid
Formel: C₇H₈Cl₃NO
MDL: MFCD01114587
CAS-Nummer: 15256-10-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR