2-Amino-4-(4-chlorphenyl)thiazol
Lieferant: BIONET RESEARCH
Synonyme:
4-(4-Chlorophenyl)thiazol-2-amine
5K-591S.500
5K-591S.1
5K-591S.5
5K-591S.10
BIOT5K-591S.500EA
0
EUR
BIOT5K-591S.500
BIOT5K-591S.1
BIOT5K-591S.5
BIOT5K-591S.10
2-Amino-4-(4-chlorphenyl)thiazol
2-Amino-4-(4-chlorophenyl)thiazole
Formel:
C₉H₇ClN₂S Molecular Weight: 210.69 g/mol |
MDL:
MFCD00047058 CAS-Nummer: 2103-99-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...