2-Chlor-5-formylpyridin
Lieferant: BIONET RESEARCH
5L-349S.500
5L-349S.1
5L-349S.5
5L-349S.10
BIOT5L-349S.500EA
0
EUR
BIOT5L-349S.500
BIOT5L-349S.1
BIOT5L-349S.5
BIOT5L-349S.10
2-Chlor-5-formylpyridin
2-Chlor-5-formylpyridin
Formel:
C₆H₄ClNO |
MDL:
MFCD03095223 CAS-Nummer: 23100-12-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...