2-Chlor-5-formylpyridin

Lieferant: BIONET RESEARCH

5L-349S.500 5L-349S.1 5L-349S.5 5L-349S.10
BIOT5L-349S.500EA 0 EUR
BIOT5L-349S.500 BIOT5L-349S.1 BIOT5L-349S.5 BIOT5L-349S.10
2-Chlor-5-formylpyridin
2-Chlor-5-formylpyridin
Formel: C₆H₄ClNO
MDL: MFCD03095223
CAS-Nummer: 23100-12-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR