2-(Methylsulfonyl)benzoesäure
Lieferant: BIONET RESEARCH
5L-603S.500
5L-603S.1
5L-603S.5
5L-603S.10
BIOT5L-603S.500EA
0
EUR
BIOT5L-603S.500
BIOT5L-603S.1
BIOT5L-603S.5
BIOT5L-603S.10
2-(Methylsulfonyl)benzoesäure
2-(Methylsulphonyl)benzoic acid
Formel:
C₈H₈O₄S |
MDL:
MFCD00216492 CAS-Nummer: 33963-55-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...