6-Brompiperonal

Lieferant: BIONET RESEARCH

Synonyme: 6-Brom-1,3-benzodioxol-5-carbaldehyd

5M-952.500 5M-952.1 5M-952.5 5M-952.10
BIOT5M-952.500EA 0 EUR
BIOT5M-952.500 BIOT5M-952.1 BIOT5M-952.5 BIOT5M-952.10
6-Brompiperonal
6-Brompiperonal
Formel: C₈H₅BrO₃
MDL: MFCD00022952
CAS-Nummer: 15930-53-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR