6-Brompiperonal
Lieferant: BIONET RESEARCH
5M-952.500
5M-952.1
5M-952.5
5M-952.10
BIOT5M-952.500EA
0
EUR
BIOT5M-952.500
BIOT5M-952.1
BIOT5M-952.5
BIOT5M-952.10
6-Brompiperonal
6-Brompiperonal
Formel:
C₈H₅BrO₃ |
MDL:
MFCD00022952 CAS-Nummer: 15930-53-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...