3-Amino-3-(4-fluorphenyl)propionsäure

Lieferant: BIONET RESEARCH

6R-0615.500 6R-0615.1 6R-0615.5
BIOT6R-0615.500EA 0 EUR
BIOT6R-0615.500 BIOT6R-0615.1 BIOT6R-0615.5
3-Amino-3-(4-fluorphenyl)propionsäure
3-Amino-3-(4-fluorphenyl)propionsäure
Formel: C₉H₁₀FNO₂
MDL: MFCD00181811
CAS-Nummer: 325-89-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR