3-Amino-3-(4-bromphenyl)propionsäure

Lieferant: BIONET RESEARCH

6R-0633.500 6R-0633.1 6R-0633.5
BIOT6R-0633.500EA 0 EUR
BIOT6R-0633.500 BIOT6R-0633.1 BIOT6R-0633.5
3-Amino-3-(4-bromphenyl)propionsäure
3-Amino-3-(4-bromphenyl)propionsäure
Formel: C₉H₁₀BrNO₂
MDL: MFCD00187112
CAS-Nummer: 39773-47-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR