6-Piperidinonicotinsäure

Lieferant: BIONET RESEARCH

Synonyme: 6-(Piperidin-1-yl)nicotinic acid, 6-(1-Piperidinyl)pyridine-3-carboxylic acid, 6-(Piperidin-1-yl)pyridine-3-carboxylic acid

6W-0335.500 6W-0335.1 6W-0335.5 6W-0335.10
BIOT6W-0335.500EA 0 EUR
BIOT6W-0335.500 BIOT6W-0335.1 BIOT6W-0335.5 CHMP6W-0335.10
6-Piperidinonicotinsäure
6-Piperidinonicotinsäure
Formel: C₁₁H₁₄N₂O₂
Molecular Weight: 206.2411 g/mol
MDL: MFCD03791231
CAS-Nummer: 120800-50-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR