2-(Chlormethyl)-5-nitrobenzoxazol
Lieferant: BIONET RESEARCH
6X-0702.500
6X-0702.1
6X-0702.5
6X-0702.10
BIOT6X-0702.500EA
0
EUR
BIOT6X-0702.500
BIOT6X-0702.1
BIOT6X-0702.5
BIOT6X-0702.10
2-(Chlormethyl)-5-nitrobenzoxazol
2-(Chloromethyl)-5-nitrobenzoxazole
Formel:
C₈H₅ClN₂O₃ |
MDL:
MFCD01596292 CAS-Nummer: 119198-10-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...