2-Amino-4-(4-bromphenyl)-5-pyrimidincarbonitril
Lieferant: BIONET RESEARCH
6Y-0837.10
6Y-0837.5
6Y-0837.1
6Y-0837.500
BIOT6Y-0837.10EA
0
EUR
BIOT6Y-0837.10
BIOT6Y-0837.5
BIOT6Y-0837.1
BIOT6Y-0837.500
2-Amino-4-(4-bromphenyl)-5-pyrimidincarbonitril
2-Amino-4-(4-bromphenyl)-5-pyrimidincarbonitril
Formel:
C₁₁H₇N₄Br |
MDL:
MFCD06659571 CAS-Nummer: 749885-93-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...