2-(6-Chlor-3-pyridazinyl)-2-(4-methylphenyl)acetonitril

Lieferant: BIONET RESEARCH

Synonyme: 6-Chloro-alpha-(4-methylphenyl)-3-pyridazineacetonitrile

7E-003.500 7E-003.1 7E-003.5 7E-003.10
BIOT7E-003.500EA 0 EUR
BIOT7E-003.500 BIOT7E-003.1 BIOT7E-003.5 BIOT7E-003.10
2-(6-Chlor-3-pyridazinyl)-2-(4-methylphenyl)acetonitril
2-(6-Chlor-3-pyridazinyl)-2-(4-methylphenyl)acetonitril
Formel: C₁₃H₁₀ClN₃
MDL: MFCD00214777
CAS-Nummer: 339008-32-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR