3-(4-Methylphenyl)-2,1-benzisoxazol-5-carbaldehyd
Lieferant: BIONET RESEARCH
7H-923.500
7H-923.1
7H-923.5
7H-923.10
BIOT7H-923.500EA
0
EUR
BIOT7H-923.500
BIOT7H-923.1
BIOT7H-923.5
BIOT7H-923.10
3-(4-Methylphenyl)-2,1-benzisoxazol-5-carbaldehyd
3-(4-Methylphenyl)-2,1-benzisoxazol-5-carbaldehyd
Formel:
C₁₅H₁₁NO₂ |
MDL:
MFCD00663843 CAS-Nummer: 328547-38-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...