3-(4-Chlorphenyl)-2,1-benzisoxazol-5-carbaldehyd

Lieferant: BIONET RESEARCH

7H-924.500 7H-924.1 7H-924.5 7H-924.10
BIOT7H-924.500EA 0 EUR
BIOT7H-924.500 BIOT7H-924.1 BIOT7H-924.5 BIOT7H-924.10
3-(4-Chlorphenyl)-2,1-benzisoxazol-5-carbaldehyd
3-(4-Chlorphenyl)-2,1-benzisoxazol-5-carbaldehyd
Formel: C₁₄H₈NO₂Cl
MDL: MFCD00663844
CAS-Nummer: 328272-55-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR