3-(4-Chlorphenyl)-2,1-benzisoxazol-5-carbaldehyd
Lieferant: BIONET RESEARCH
7H-924.500
7H-924.1
7H-924.5
7H-924.10
BIOT7H-924.500EA
0
EUR
BIOT7H-924.500
BIOT7H-924.1
BIOT7H-924.5
BIOT7H-924.10
3-(4-Chlorphenyl)-2,1-benzisoxazol-5-carbaldehyd
3-(4-Chlorphenyl)-2,1-benzisoxazol-5-carbaldehyd
Formel:
C₁₄H₈NO₂Cl |
MDL:
MFCD00663844 CAS-Nummer: 328272-55-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...