O-(4-Nitrobenzyl)hydroxylamin Hydrochlorid

Lieferant: BIONET RESEARCH

Synonyme: O-(4-Nitrobenzyl)-hydroxylammonium chloride

7L-014.500 7L-014.1 7L-014.5 7L-014.10
BIOT7L-014.500EA 0 EUR
BIOT7L-014.500 BIOT7L-014.1 BIOT7L-014.5 BIOT7L-014.10
O-(4-Nitrobenzyl)hydroxylamin Hydrochlorid
O-(4-Nitrobenzyl)hydroxylamin Hydrochlorid
Formel: C₇H₉ClN₂O₃
Molecular Weight: 204.61 g/mol
MDL: MFCD00012954
CAS-Nummer: 2086-26-2
EINECS: 218-228-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR