4,5-Dibrom-3(2H)-pyridazinon
Lieferant: BIONET RESEARCH
Synonyme:
4,5-Dibromopyridazin-3(2H)-one
7M-627S.500
7M-627S.1
7M-627S.5
7M-627S.10
BIOT7M-627S.500EA
0
EUR
BIOT7M-627S.500
BIOT7M-627S.1
BIOT7M-627S.5
BIOT7M-627S.10
4,5-Dibrom-3(2H)-pyridazinon
4,5-Dibrom-3(2H)-pyridazinon
Formel:
C₄H₂Br₂N₂O Molecular Weight: 253.89 g/mol |
MDL:
MFCD00023641 CAS-Nummer: 5788-58-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...