3-Amino-3-(3-bromphenyl)propionsäure

Lieferant: BIONET RESEARCH

Synonyme: DL-3-Amino-3-(3-Bromophenyl)propionic acid

7R-0609.500 7R-0609.1 7R-0609.5
BIOT7R-0609.500EA 0 EUR
BIOT7R-0609.500 BIOT7R-0609.1 BIOT7R-0609.5
3-Amino-3-(3-bromphenyl)propionsäure
3-Amino-3-(3-bromphenyl)propionsäure
Formel: C₉H₁₀BrNO₂
Molecular Weight: 244.0852 g/mol
MDL: MFCD00995843
CAS-Nummer: 117391-50-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR