3-Amino-3-(3-methoxyphenyl)propionsäure

Lieferant: BIONET RESEARCH

Synonyme: 3-Amino-3-(3-methoxyphenyl)propanoic acid

7R-0632.500
BIOT7R-0632.500EA 0 EUR
BIOT7R-0632.500
3-Amino-3-(3-methoxyphenyl)propionsäure
3-Amino-3-(3-methoxyphenyl)propionsäure
Formel: C₁₀H₁₃NO₃
Molecular Weight: 195,22 g/mol
MDL: MFCD00462569
CAS-Nummer: 68208-19-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR