2-Chlor-4-fluor-5-nitroanisol

Lieferant: BIONET RESEARCH

7T-0245.500 7T-0245.1 7T-0245.5 7T-0245.10
BIOT7T-0245.500EA 0 EUR
BIOT7T-0245.500 BIOT7T-0245.1 BIOT7T-0245.5 BIOT7T-0245.10
2-Chlor-4-fluor-5-nitroanisol
2-Chlor-4-fluor-5-nitroanisol
Formel: C₇H₅ClFNO₃
MDL: MFCD03425810
CAS-Nummer: 84478-76-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR