N-(4-Nitrobenzoyl)morpholin

Lieferant: BIONET RESEARCH

Synonyme: Morpholino(4-nitrophenyl)methanone

7X-0701.500 7X-0701.1 7X-0701.5 7X-0701.10
BIOT7X-0701.500EA 0 EUR
BIOT7X-0701.500 BIOT7X-0701.1 BIOT7X-0701.5 BIOT7X-0701.10
N-(4-Nitrobenzoyl)morpholin
N-(4-Nitrobenzoyl)morpholin
Formel: C₁₁H₁₂N₂O₄
Molecular Weight: 236.23 g/mol
MDL: MFCD00451121
CAS-Nummer: 5397-76-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR