6-Chlorimidazo[1,2-a]pyridin

Lieferant: BIONET RESEARCH

7X-0892.500 7X-0892.1 7X-0892.5 7X-0892.10
BIOT7X-0892.500EA 0 EUR
BIOT7X-0892.500 BIOT7X-0892.1 BIOT7X-0892.5 BIOT7X-0892.10
6-Chlorimidazo[1,2-a]pyridin
6-Chlorimidazo[1,2-a]pyridin
Formel: C₇H₅ClN₂
MDL: MFCD05663807
CAS-Nummer: 6188-25-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR