6-Bromimidazo[1,2-a]pyridin

Lieferant: BIONET RESEARCH

7X-0903.1 7X-0903.10 7X-0903.5 7X-0903.500
BIOT7X-0903.1EA 0 EUR
BIOT7X-0903.1 BIOT7X-0903.10 BIOT7X-0903.5 BIOT7X-0903.500
6-Bromimidazo[1,2-a]pyridin
6-Bromimidazo[1,2-a]pyridin
Formel: C₇H₅BrN₂
MDL: MFCD06496200
CAS-Nummer: 6188-23-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR