3-Amino-3-(3-nitrophenyl)propionsäure

Lieferant: BIONET RESEARCH

8R-0617.500 8R-0617.1 8R-0617.5
BIOT8R-0617.500EA 0 EUR
BIOT8R-0617.500 BIOT8R-0617.1 BIOT8R-0617.5
3-Amino-3-(3-nitrophenyl)propionsäure
3-Amino-3-(3-nitrophenyl)propionsäure
Formel: C₉H₁₀N₂O₄
MDL: MFCD00472612
CAS-Nummer: 5678-47-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR