3-Amino-3-(4-hydroxyphenyl)propionsäure

Lieferant: BIONET RESEARCH

Synonyme: 3-Amino-3-(4-hydroxyphenyl)propanoic Acid

8R-0644.500 8R-0644.1 8R-0644.5
BIOT8R-0644.500EA 0 EUR
BIOT8R-0644.500 BIOT8R-0644.1 BIOT8R-0644.5
3-Amino-3-(4-hydroxyphenyl)propionsäure
3-Amino-3-(4-hydroxyphenyl)propionsäure
Formel: C₉H₁₁NO₃
MDL: MFCD00181810
CAS-Nummer: 6049-54-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR