3-Amino-4-ethylcapronsäure

Lieferant: BIONET RESEARCH

8T-0051.500 8T-0051.1 8T-0051.5 8T-0051.10
BIOT8T-0051.500EA 0 EUR
BIOT8T-0051.500 BIOT8T-0051.1 BIOT8T-0051.5 BIOT8T-0051.10
3-Amino-4-ethylcapronsäure
3-Amino-4-ethylcapronsäure
Formel: C₈H₁₇NO₂
Molecular Weight: 159.23 g/mol
MDL: MFCD01863280
CAS-Nummer: 204191-42-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR