Lanthan(III)fluorid
Lieferant: BIONET RESEARCH
Synonyme:
Lanthanum trifluoride
902405-0100
BIOT902405-0100EA
0
EUR
BIOT902405-0100
Lanthan(III)fluorid
Lanthan(III)fluorid
Formel:
F₃La Molecular Weight: 195,9 g/mol Siedepunkt: ∼2327 °C (1013 hPa) Schmelzpunkt: 1491,85…1507.85 °C Dichte: 5,936 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011073 CAS-Nummer: 13709-38-1 EINECS: 237-252-8 UN: 3288 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...