3-(Boc-amino)-3-(4-methoxyphenyl)propionsäure

Lieferant: BIONET RESEARCH

9R-0657.500 9R-0657.1
BIOT9R-0657.500EA 0 EUR
BIOT9R-0657.500 BIOT9R-0657.1
3-(Boc-amino)-3-(4-methoxyphenyl)propionsäure
3-(Boc-amino)-3-(4-methoxyphenyl)propionsäure
Formel: C₁₅H₂₁NO₄
MDL: MFCD02090694
CAS-Nummer: 96363-20-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR