6-Chlor-4-nitro-o-toluidin
Lieferant: BIONET RESEARCH
|
Gefahr
|
Synonyme:
2-Chlor-6-methyl-4-nitroanilin, 2-Amino-3-chlor-5-nitrotoluol, 6-Chlor-2-methyl-4-nitroanilin
9T-0036.500
9T-0036.1
9T-0036.5
9T-0036.10
BIOT9T-0036.500EA
0
EUR
BIOT9T-0036.500
BIOT9T-0036.1
BIOT9T-0036.5
BIOT9T-0036.10
6-Chlor-4-nitro-o-toluidin
6-Chlor-4-nitro-o-toluidin
Formel:
C₇H₇ClN₂O₂ Schmelzpunkt: 172,5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01313703 CAS-Nummer: 69951-02-6 UN: 2237 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...