4'-(Methylsulfonyl)acetophenon

Lieferant: BIONET RESEARCH

9W-0290.500 9W-0290.1 9W-0290.5 9W-0290.10
BIOT9W-0290.500EA 0 EUR
BIOT9W-0290.500 BIOT9W-0290.1 BIOT9W-0290.5 BIOT9W-0290.10
4'-(Methylsulfonyl)acetophenon
4'-(Methylsulfonyl)acetophenon
Formel: C₉H₁₀O₃S
MDL: MFCD00025072
CAS-Nummer: 10297-73-1
EINECS: 233-672-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR