2-Brom-4'-(methylsulfonyl)acetophenon

Lieferant: BIONET RESEARCH

9W-0291.500 9W-0291.1 9W-0291.5 9W-0291.10
BIOT9W-0291.500EA 0 EUR
BIOT9W-0291.500 BIOT9W-0291.1 BIOT9W-0291.5 BIOT9W-0291.10
2-Brom-4'-(methylsulfonyl)acetophenon
Formel: C₉H₉BrO₃S
MDL: MFCD00673134
CAS-Nummer: 50413-24-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR