2-Brom-4'-(methylsulfonyl)acetophenon
Lieferant: BIONET RESEARCH
9W-0291.500
9W-0291.1
9W-0291.5
9W-0291.10
BIOT9W-0291.500EA
0
EUR
BIOT9W-0291.500
BIOT9W-0291.1
BIOT9W-0291.5
BIOT9W-0291.10
2-Brom-4'-(methylsulfonyl)acetophenon
Formel:
C₉H₉BrO₃S |
MDL:
MFCD00673134 CAS-Nummer: 50413-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...