2-Piperidinoisonicotinonitril

Lieferant: BIONET RESEARCH

9X-0702.1 9X-0702.10 9X-0702.5 9X-0702.500
BIOT9X-0702.1EA 0 EUR
BIOT9X-0702.1 BIOT9X-0702.10 BIOT9X-0702.5 BIOT9X-0702.500
2-Piperidinoisonicotinonitril
2-Piperidinoisonicotinonitril
Formel: C₁₁H₁₃N₃
MDL: MFCD06200992
CAS-Nummer: 127680-89-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR