3-Fluor-4-nitrobenzaldehyd

Lieferant: BIONET RESEARCH

FA-0630.10
BIOTFA-0630.10EA 0 EUR
BIOTFA-0630.10
3-Fluor-4-nitrobenzaldehyd
3-Fluoro-4-nitrobenzaldehyde
Formel: C₇H₄NO₃F
Molecular Weight: 169.1100 g/mol
MDL: MFCD00968940
CAS-Nummer: 160538-51-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR